Axygen® Conical-Bottom Centrifuge Tubes, Polypropylene, Corning
Supplier: Corning
SCT-15ML-500 SCT-50ML-R-S SCT-15ML-25-S SCT-50ML-25-S SCT-50ML-500
89005-586CS 195.53 CAD
89005-586 89005-588 89005-622 89005-624 89005-592
Axygen® Conical-Bottom Centrifuge Tubes, Polypropylene, Corning
Tubes Centrifuge Tubes
Polypropylene tubes feature printed graduations and a writing area for sample identification.
  • Caps may be closed with one hand
  • Tubes are autoclavable
The 15 ml tubes can be centrifuged to 7500 g; the 50 ml tubes can be centrifuged to 9400 g.