VACUETTE® Safety Blood Collection Sets, Greiner Bio-One
Supplier: Greiner bio-one
Vacuette®
450086 450090 450095 450161 450160 450081 450092 450082 450085 450099 450096 450091
10799-060CS 640.88 CAD
10799-060 95057-317 10861-118 95057-315 95057-319 95057-321 95057-331 10861-120 10799-058 89370-872 95057-325 95057-323
VACUETTE® Safety Blood Collection Sets, Greiner Bio-One
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Blood collection sets are designed to increase the safety of routine venipuncture procedures.
  • Single-use, sterile, winged blood collection needles bonded to flexible tubing
  • Protective shield helps prevent needlestick injuries
  • Off-center Luer needle adapter
  • Flat venipuncture angle
The completely assembled QUICKSHIELD safety tube holders are equipped with a rotational protective shield that can be activated with the aid of a solid surface
Sets consist of a single-use, sterile, winged blood collection needle bonded to flexible tubing. The winged needle is designed with a safety shield, which can be activated to cover the needle immediately following blood collection to aid in the protection against accidental needlestick injury. Sets are available with or without Luer adapter, with Luer adapter and holder, or with Luer adapter and HOLDEX® holder. The HOLDEX® standard holder is distinguished by its off-center Luer needle adapter, which provides a flat venipuncture angle.
The protective shield is attached to the holder and, once activated, keeps the needle contained to help prevent needlestick injuries. The rotational shield can rotate 360° to provide user comfort. Holders are designed exclusively for use with VACUETTE® multiple sample drawing needles or VACUETTE® VISIO PLUS needles. The QUICKSHIELD Complete combines a safety tube holder and a VACUETTE® multiple sample drawing needle. The QUICKSHIELD Complete PLUS combines a safety tube holder and a VACUETTE® VISIO PLUS needle.