VACUETTE® EDTA Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
454021 456038 456003 455036 454428 456002 456058P 454222 454217 456067 456074 454246 454209 454208 456279
95057-225CS 386.93 CAD
95057-225 95057-227 95057-229 95057-231 95057-237 95057-239 95057-247 95057-251 95057-253 95057-221 95057-223 95057-233 95057-235 95057-241 95057-243 95057-255 95057-245 75800-314
VACUETTE® EDTA Tubes, Greiner Bio-One
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Designed for use in the examination of whole blood in hematology applications, EDTA tubes feature interior walls that are coated with either K2EDTA or K3EDTA.
  • Gel separator forms stable barrier between serum and blood cells
  • PET plastic construction is virtually unbreakable
  • Color-coded rings in caps
The EDTA binds calcium ions and blocks the coagualtion cascade
The EDTA tubes equipped with a gel separator contain a barrier gel in the base of the tube. During centrifugation, this gel forms a stable barrier between the serum and the blood cells,More Product Information