KIMAX® Centrifuge Tubes with [ST] Glass Stopper, Conical-Bottom, Graduated, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
45153-15 45153-50
89002-184CS 1240.17 CAD
89002-184 89002-186
KIMAX® Centrifuge Tubes with [ST] Glass Stopper, Conical-Bottom, Graduated, Kimble Chase
Tubes Centrifuge Tubes
Tubes feature solid, pennyhead [ST] stoppers.
  • Graduated and calibrated to contain
  • Ext.Ø and Length do not include stopper
Scale, legend, and marking spot are printed in durable white ceramic enamel. The 15 ml tube is graduated in 0.1 ml increments. The 50 ml tube is graduated to 10 ml in 0.5 ml increments, and from 10 to 50 ml in 1.0 ml increments.
Caution: Do not centrifuge with stopper in tube.