Nalgene® Round-Bottom Centrifuge Tubes, Polypropylene Copolymer, Thermo Scientific
Supplier: Thermo Fisher Scientific
3110-0120 3110-0150 3110-0160 3110-0380 3110-9500 3110-0500 3110-1000
21010-749CS 1214.3 CAD
21010-749 21010-760 21010-771 21010-807 21010-818 21010-829 21010-830
Nalgene® Round-Bottom Centrifuge Tubes, Polypropylene Copolymer, Thermo Scientific
Tubes Centrifuge Tubes
Heavy-duty construction for repeated autoclavability.
  • Excellent chemical and abrasion resistance
  • An economical choice for refrigerated or standard centrifugation
Note: For proper performance, fill tubes to at least 80% of the total capacity.
Molded for uniform wall thickness and extra strength. PPCO has a better low-temperature rating than standard polypropylene. Also useful as test tubes. RCF ratings are based on chemical compatibility and proper rotor fit.
Ordering information: Supplied without closures. Polypropylene closures availableMore Product Information