PYREX® Heavy Duty Round Bottom Centrifuge Tubes, Corning
Supplier: Corning
8422-100 8422-50
21023-403CS 131.39 CAD
21023-403 89091-460
PYREX® Heavy Duty Round Bottom Centrifuge Tubes, Corning
Tubes Centrifuge Tubes
PYREX®, borosilicate glass, round bottom, with screw cap.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These 50 ml round bottom centrifuge tubes are useful for the handling of sputum specimens. Digestion, shaking, neutralizing and centrifuging can be performed in one container. They are also useful for other clinical and microbiological tests. Screw caps resist the effects of temperature and steam. The 50 ml tube has a black phenolic GPI 24-410 threaded cap with a glued-in, white rubber liner.