Sieves, Brass Wire and Frame, W.S. Tyler™
Supplier: W.S. Tyler
4529 4530 4531 4532 4533 4534 4535 4536 4537 4538 4539 4543 4544 4545 4546 4547 4548 4549 4550 4551 4552 4695 4696 4697 4699 4700 4701 4702 4703 4704 4705 4706 4707 4708 4710 4711 4712 4713 4714 4715 4717 4718 4541 4542
57320-486EA 439.37 CAD
57320-486 57320-500 57320-522 57320-544 57320-566 57320-588 57320-602 57320-624 57320-646 57320-668 57320-680 57320-760 57320-781 57320-806 57320-828 57320-840 57320-861 57320-883 57320-908 57320-920 57320-941 57331-405 57331-427 57331-449 57331-482 57331-507 57331-529 57331-540 57331-562 57331-584 57331-609 57331-620 57331-642 57331-664 57331-700 57331-722 57331-744 57331-766 57331-788 57331-802 57331-846 57331-868 57320-726 57320-748
Sieves, Brass Wire and Frame, W.S. Tyler™
Sieves Test Sieves
Full height sieves are 5.08cm (2") high above brass wire cloth
Each sieve is serialized and supplied with a “Test Sieve Certificate.” Sieves are marked with the USA Series Number. Sieves one millimeter and coarser are identified by opening in millimeters and inches; sieves finer, by opening in microns and inches.
Overall height: 7.4cm (2.9"). Half height sieves are 2.54cm (1") high above brass wire cloth. Overall height: 4.8cm (115/16"). Diameter: 20.3cm (8").
Certifications: Conform to ASTM E-11 specifications.