Orion™ pH Buffer Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
910760
910460
911060
910104
910105
910107
910110
910168
910686
910918
916860
910112
9116860
9191860
23198-220EA
99.69
CAD
23198-220
23198-222
23198-224
34182-306
34182-310
34182-386
34182-390
CA10010-636
CA10010-638
CA10010-640
CA10010-644
CA10010-634
CA10010-646
CA10010-648
Orion™ pH Buffer Bottles, Thermo Scientific
Buffers
pH Calibration Buffers
Perform accurate and reproducible pH calibrations with Orion™ pH buffer bottles.
- All bottles feature pH/temperature tables printed on the labels for accurate calibrations and NIST traceability
- Manufactured under ISO 9000 quality standards
- Available in pint (475 ml) bottles or portable five packs of 2 oz. (5×60 ml) bottles
The pH 4.01, 5.00, 7.00, and 10.01 buffers are color-coded for ease of use. All Orion™ pH buffers are traceable to NIST standard reference materials.