pH Buffer Capsules, Hydrion®
Supplier: MICRO ESSENTIAL LABORATORY I
270-2.00 270-2.40 270-3.00 270-4.00 270-5.00 270-6.80 270-7.00 270-7.20 270-7.40 270-8.00 270-8.40 270-9.00 270-10.00 270-11.00 270-12.00
34175-027CS 114.79 CAD
34175-027 34175-049 34175-071 34175-129 34175-173 34175-231 34175-242 34175-297 34175-300 34175-322 34175-344 34175-377 34175-399 34175-435 34175-480
pH Buffer Capsules, Hydrion®
Buffers pH Calibration Buffers
For the quick and convenient preparation of fresh solutions of known pH values
Capsule contents dissolve in 100mL of distilled water, giving a clear solution accurate to ±0.02 pH unit. The dry powder remains stable. Each vial contains 10 capsules.