J.T.Baker®, pH Buffer Solutions
Supplier: Avantor
J.T.Baker®
BAKR5606-01 5655-03 BAKR5655-03 5606-03 5657-07 5655-01 5655-07 5656-07 5657-03 BAKR5657-07 BAKR5606-03 BAKR5655-01 BAKR5657-01 BAKR5657-03 BAKR5608-01 BAKR5657-02 5606-01 BAKR5608-03 5657-01 5656-01
CAJT5657-7EA 672.78 CAD
40004-722 CAJT5657-7 CAJT5606-1 CAJT5656-7 JT5655-1 CAJT5657-3 CAJT5606-3 CAJT5655-7 CAJT5655-1 CAJT5655-3 JT5657-7 CAJT5657-1 CAJT5656-1 JT5655-3 JT5657-1 JT5606-3 JT5608-1 JT5657-3 JT5606-1 JT5608-3
J.T.Baker®, pH Buffer Solutions
Buffers pH Calibration Buffers
High quality buffer solutions ensure accurate pH measurements time after time.
  • Colour coded
  • Potassium hydrogen phthalate based pH 4 buffer (red)
  • Phosphate based pH 7 buffer (yellow)
  • Borate based pH 10 buffer (blue)
Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilize quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardization of solutions, helping streamline high-volume analytical work in applications such as quality control.
Solutions are available in both clear and color-coded varieties.
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Certifications: Certified traceable to NIST Standard Reference Buffers.