Macron Fine Chemicals™ BuffAR®, pH Buffer Solutions
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MACR0031-04
MACR0031-10
MACR0097-10
MACR0098-04
MACR0098-10
MACR0099-10
MACR0029-04
MACR0097-04
MACR0099-04
CAMK002904CS
1096.76
CAD
CAMK002904
CAMK003104
CAMK003110
CAMK009710
CAMK009804
CAMK009810
CAMK009910
MK0029-04
CAMK009704
CAMK009904
Macron Fine Chemicals™ BuffAR®, pH Buffer Solutions
Buffers
pH Calibration Buffers
Accurate to within ±0.01 pH at 25 °C (±0.02 for pH 9 and 9.18).
Available as clear or color-coded solutions.