BRAND Centrifuge Tube Mini-Coolers, BrandTech
Supplier: Brandtech
114935 114940 114930
89048-942EA 658.25 CAD
89048-942 89048-944 89048-948
BRAND Centrifuge Tube Mini-Coolers, BrandTech
Racks Tube Racks
Centrifuge tube mini-coolers accommodate twelve microtubes ranging from 0.5 to 2.0 ml.
  • Maintain temperature of microcentrfuge tubes
  • Transparent lid provides visual confirmation of contents
  • For benchtop, or in-lab transport
When pre-frozen, an insulating gel allows mini-coolers to maintain the rated temperature for approximately one hour (45 minutes for –70°C model). Each model is supplied with a transparent lid.