Alundum® Extraction Thimbles
Supplier: Saint Gobain Performance Plastics
3254422 3254422
27720-124EA 50.52 CAD
27720-124 27721-047 27721-069 27721-149 27721-068
Alundum® Extraction Thimbles
Extractors Extraction Thimbles
Thimbles remain very constant in weight
Preferable to paper thimbles as they may be used an indefinite number of times and permit the ignition of the extracted residue without removing the material from the thimble. To clean, place in a strong solution of acid for a short time and then ignite to constant weight in the usual way. Available in three degrees of porosity.