PYREX® Extraction Thimbles, with Fritted Disc, Corning
Supplier: Corning
33950-MEC 33950-SC 33950-MC 33950-LC 33950-LEC
27743-120CS 213.11 CAD
27743-120 89091-272 89091-346 89091-384 27743-141
PYREX® Extraction Thimbles, with Fritted Disc, Corning
Extractors Extraction Thimbles
PYREX®, borosilicate glass, with fritted disc.
  • Resistant to chemical attack
  • Excellent temperature resistance
It is designed for use with soxhlet extractors. It is also useful for chemical applications where it is required to weigh the extracted material.