Rifampicin, Calbiochem®

Supplier: MilliporeSigma

557303-1GM 557303-5GM
CA80058-624EA 309.78 CAD
CA80058-624 CA80108-288
Rifampicin, Calbiochem®
Rifampicin

Rifampicin is an antibiotic that specifically inhibits DNA-dependent RNA polymerase in bacteria by forming an inactive complex. Does not affect mammalian RNA polymerase. Inhibits transcription by preventing the initial transcription complex from entering the elongation mode.

Formula: C₄₃H₅₈N₄O₁₂
Melting Pt: 183…188 °C
MDL Number: MFCD00151389
CAS Number: 13292-46-1
EINECS: 236-312-0
Merck Index: 12,08382

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR