VWR® Centrifuge Tubes, Polystyrene
Supplier: VWR
VWR®
CFT459150 CFT429150 CFT415150 CFT428150 CFT430150
21008-202CS 504.19 CAD
21008-202 21008-187 21008-212 21008-189 21008-197
VWR® Centrifuge Tubes, Polystyrene
Tubes Centrifuge Tubes
These tubes are made of optically clear PS and feature distinct molded graduations.
  • Colorless and transparent are perfect for biological reaction experiments
  • RNase-, DNase-, and Endotoxin-Free
  • Leak-proof
  • Not autoclavable
  • Sterile tubes are sterilized by irradiation
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
The flat surface of the polypropylene cap provides an at-a-glance identification area.
Certifications: Manufactured in a class 100000 room environment. Manufactured under ISO13485:2016 and ISO9001:2015 quality management. Meet the requirements of RoHS.
Ordering information: Choose from tubes and caps supplied in Earth Friendly cardboard racks, bag packed, or bulk packed.