Deluxe Pallet Trucks, Wesco
Supplier: Wesco
272765 272748
21002-052EA 2931.96 CAD
21002-052 21002-054
Deluxe Pallet Trucks, Wesco
Trolleys
Designed for economical, convenient lifting, transporting, and lowering of wooden pallets.
  • Pump overload bypass
  • Rubber-coated safety loop handle features a triple function hand control (raise, neutral, lower) to maneuver easily in any position
  • Adjustable fork tie rods
  • Tapered fork tips with rugged entry guides help reduce damage
  • Box-formed, 8-gauge, 15.2 cm (6") wide, high-tensile steel forks are structurally reinforced at critical stress points to help prevent bending
  • Large mold-on polyurethane steering wheel allows smooth rolling over uneven surfaces
  • A 200° steering angle allows extra maneuverability in tight areas
  • Exit rollers
Fork footprint: 27x48"
Ordering information: Manufacturer's five-year warranty on pump.