PIG® Universal Pads and Rolls, New Pig
Supplier: New Pig
MAT204 MAT230
56620-034PK 299.09 CAD
56620-034 56620-040
PIG® Universal Pads and Rolls, New Pig
Spill Control Materials
Polypropylene absorbent pads and rolls are designed to safeguard walking areas against liquids that could cause accidents and for daily protection in areas prone to overspray or sporadic drips, such as under conveyors.
  • Eight-layer pads and rolls combine grip and strength to reduce slip hazards and keep floors clean
  • Polypropylene (made with 50% recycled content) soaks up and retains liquids
  • Dimpled for quicker absorption of water-based liquids, petroleum products, solvents, and a broad range of chemicals
  • Choose light, medium, or heavyweight depending on the amount of liquid
  • Can be incinerated after use to reduce waste
Dark gray material is dimpled for quicker absorption of water-based liquids, petroleum products, solvents, and a broad range of chemicals. Lightweight pads and rolls are flexible for wiping up small leaks and lining tool boxes and benches. Mediumweight pads and rolls are versatile for general use. Heavyweight pads and rolls are constructed for large-volume cleanups. Rolls are perforated every 25.4cm (10") for tearing off the length of pad needed. Can be wrung out and incinerated after use to reduce waste.
Note: Given capacity is absorbency capacity of entire package.