BD Clay Adams™ SurePrep™ Capillary Tubes, BD Medical
Supplier: BD
420314 420316
CA15401-562PK 298.91 CAD
CA15401-562 CA15401-566
BD Clay Adams™ SurePrep™ Capillary Tubes, BD Medical
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Unlike conventional sealing capillary tubes, SurePrep™ tubes seal automatically when the blood sample touches the self-sealing plug
In the event of an accidental break, the protective Mylar® wrap helps contain blood and minimizes exposure to broken glass.
Ordering information: Packed 100 tubes per vial.