Thiostrepton, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: N-[(7R,8S)-2-Carboxy-7,8-dihydro-8-hydroxy-4-[(1S)-1-hydroxyethyl]-7-quinolinyl]-L-isoleucyl-L-alanyl-2,3-didehydroalanyl-L-alanyl-2-[(4aR,11S,14Z,18S,21S,28S,32aS)-4a-amino-21-[(1S,2R)-1,2-dihydroxy-1-methylpropyl]-14-ethylidene-3,4,4a,9,10,11,12,13,14,18,19,20,21,27,28,32a-hexadecahydro-11,28-bis[(1R)-1-hydroxyethyl]-9,12,19,26-tetraoxo-17H,26H-8,5:18,15:25,22:32,29-tetranitrilo-5H,15H-pyrido[3,2-m][1,11,17,24,4,7,20,27]tetrathiatetraazacyclotriacontin-2-yl]-4-thiazolecarbonyl-2,3-didehydroalanyl-2,3-didehydro-alaninamide,(1→528)-lactone

598226-10GM 598226-1GM
CA80057-598EA 2245.84 CAD
CA80057-598 CA80057-600
Thiostrepton, Sigma-Aldrich®
Thiostrepton

A thiazole-containing peptide antibiotic that inhibits protein synthesis by preventing binding of GTP to 50S ribosomal subunit


Inhibits the function of elongation factor G (EF-G) and the dissociation of EF-G from the ribosome. The thiostrepton resistant gene is also commonly used as a selective marker for recombinant DNA/plasmid technologies. Bioassay: >=1000units/mg (zonal plate clearing assay).

Formula: C₇₂H₈₅N₁₉O₁₈S₅
MW: 1664.887 g/mol
MDL Number: MFCD00135828
CAS Number: 1393-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR