Streptomycin sulfate (from Streptomyces species), Calbiochem®

Supplier: MilliporeSigma

5711-100GM 5711-100GM
CA80058-670EA 379.84 CAD
CA80058-670 80058-670
Streptomycin sulfate (from Streptomyces species), Calbiochem®
Streptomycin sulphate

Antibiotic effective against gram-positive and gram-negative bacteria. Often used in culture media to control growth of microorganisms. Inhibits initiation, elongation and termination of protein synthesis in prokaryotes and induces misreading of the genetic code. Assay: >=700units/mg. Suggested working concentration: 25–50µg/mL.

Formula: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
MW: 1457.4 g/mol
Melting Pt: 250 °C
MDL Number: MFCD00037023
CAS Number: 3810-74-0
UN: 2811
ADR: 6.1,III
Merck Index: 12,08983

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR