Kanamycin sulfate (from Streptomyces kanamyceticus), Calbiochem®

Supplier: MilliporeSigma

420311-5GM 420311-25GM
CA80058-286EA 247.41 CAD
CA80058-286 CA80108-166
Kanamycin sulfate (from Streptomyces kanamyceticus), Calbiochem®
Kanamycin sulphate

Contains more than 98% kanamycin A.


An aminoglycoside antibiotic effective against gram-positive and gram-negative organisms. Inhibitor of protein biosynthesis that acts on the 30S ribosome, causing misreading of the genetic code. Activity ≥700 µg/mg. Recommended working concentration: 100 µg/ml.

Formula: C₁₈H₃₈N₄O₁₅S
Storage Temperature: Ambient
MDL Number: MFCD00070253
CAS Number: 25389-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR