Chloramphenicol ≥97%, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

220551-100GM 220551-500GM
CA80057-928EA 521.04 CAD
CA80057-928 CA80108-080 CA80108-082
Chloramphenicol ≥97%, Sigma-Aldrich®
Chloramphenicol

Synthetic bacteriostatic antibiotic that inhibits the translation of RNA by blocking the peptidyltransferase reaction on ribosomes.

Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
Boiling Pt: 645 °C (1013 hPa)
Melting Pt: 150 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078159
CAS Number: 56-75-7
EINECS: 200-287-4
Merck Index: 13,02087

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR