Ampicillin sodium salt, Calbiochem®

Supplier: MilliporeSigma

171254-5GM 171254-25GM
CA80055-786EA 267.95 CAD
CA80055-786 CA80108-050
Ampicillin sodium salt, Calbiochem®
Ampicillin sodium salt

Kills growing cells by interfering with the terminal reaction in bacterial wall synthesis


Active against gram-negative bacteria. Inactivated by lactamases. Suitable for use in research that uses ampicillin-resistant plasmids.

Formula: C₁₆H₁₈N₃NaO₄S
MDL Number: MFCD00064313
CAS Number: 69-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR