Mycophenolic acid ≥95% (by HPLC), Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: (E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic acid

475913-100MG 475913-500MG
CA80058-444EA 357.32 CAD
CA80058-444 CA80058-446
Mycophenolic acid ≥95% (by HPLC), Sigma-Aldrich®
Mycophenolic acid

White to off-white solid


An immunosuppressive agent that inhibits de novo purine nucleotide synthesis via inhibition of IMP dehydrogenase and prevents the formation of XMP and GMP. Useful for selection of animal cells that express the E. coli gene coding for xanthine-guanine phosphoribosyltransferase. Also suppresses cytokine-induced nitric oxide production in mouse and rat vascular endothelial cells.


Soluble in EtOH or 0.1 N NaOH

Formula: C₁₇H₂₀O₆
MW: 320.34 g/mol
MDL Number: MFCD00036814
CAS Number: 24280-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR