Tissue Freezing Media (TFM™), TBS®, General Data
Supplier: General Data Healthcare
TFM-Y TFM-R TFM-B TFM-C
CA15146-021CS 800.22 CAD
CA15146-021 CA15146-023 CA15146-025 CA15146-027 CA27900-246
Tissue Freezing Media (TFM™), TBS®, General Data
Cell Freezing Media
This fast-freezing, cryo-embedding matrix improves specimen handling
Unique formula reduces frozen specimen curling and simplifies pick-up of flat serial sections. Reduced water content minimizes freeze-fractures. Color variety makes differentiating multiple specimens easy. Size: 118mL (4oz.).
Ordering information: Assortment 15146-019 includes one bottle of each color in a wooden rack.