HyClone™ Dulbecco's Phosphate Buffered Saline, Powder, HyClone products (Cytiva)

Supplier: Cytiva

SH30013.02 SH30013.03 SH30013.04
CA16777-103EA 36.06 CAD
CA16777-103 CA16777-102 CA16777-105
HyClone™ Dulbecco's Phosphate Buffered Saline, Powder, HyClone products (Cytiva)
Buffers Cell Culture Buffers

Balanced salt solution used for multiple cell culture applications (powder form).


  • Formulated and processed for rapid dissolution
  • Non sterile
  • Pre-weighed powders ready for hydration to 1X
  • Available in 3" ported funnel shaped containers upon request
  • Available in lot sizes up to 6500 kg.


For making isotonic solutions for cell or tissue rinsing, cell dilution, reagent prep, transportation etc. where maintanence of cell integrity is required for further manufacturing or research use.


Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.


Caution: Not for diagnostic or therapeutic use.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR