Corning® Matrigel® Basement Membrane Matrix, Corning

Supplier: Corning

354262 354263 354234
CABD354262EA 1100.01 CAD
CABD354262 CABD354263 CACB354234
Corning® Matrigel® Basement Membrane Matrix, Corning
Cell Culture Supplements and Additives

Growth Factor Reduced (GFR) formulation is well suited for applications that require a more highly defined basement membrane preparation


  • Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
  • Extensive PCR testing is performed to screen for a number of pathogens
  • Tested and found negative for bacteria, fungi, and mycoplasma
  • Protein concentrations determined by Lowry method
  • Endotoxin units measured by Limulus Amoebocyte Lysate assay
  • Matrigel Matrix gel stability tested for a period of 14 days at 37°C
  • Biological activity determined for each lot using a neurite outgrowth assay.


Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Application
• Cell Growth and Differentiation
• Metabolism/Toxicology Studies
• Invasion Assays
• In Vitro and In Vivo Angiogenesis Assays
• In Vivo Angiogenesis Studies and Augmentation of Tumors in Immunosuppressed Mice

Preparation and Storage
Store at -20°C. Avoid multiple freeze-thaws


Note: For research use only. Not intended for diagnostic or therapeutic procedures.


When warmed, the solution polymerizes to produce a biologically active matrix material resembling the mammalian cellular basement membrane. It is ideal for use with in vitro cell cultures of a broad range of cell types, including both primary and stem cell applications.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR