Safety Face Shield, Sellstrom®
Supplier: Sellstrom
S39010 S35000
33007-060EA 132.02 CAD
33007-060 33007-071
Safety Face Shield, Sellstrom®
Face Shields
Provides facial protection from splashes and flying particles.
  • Economical, lightweight design
  • one-piece, molded headgear with free-floating suspension
  • Two snap-in locking headgear
  • Straps adjust independently to ensure a tight fit
Replaceable clear acetate window: 1 mm (0.04") thick
WxH: 8x12"
Caution: Note: Face shield should be worn over ANSI-approved protective eyewear.