HyClone™ Ham's Nutrient Mixture F10/F12, Cell Culture Media, Powdered, Cytiva
Supplier: Cytiva
SH30010.03 SH30010.04
CA16777-089EA 100.9 CAD
CA16777-089 CA16777-090 CA16777-092
HyClone™ Ham's Nutrient Mixture F10/F12, Cell Culture Media, Powdered, Cytiva
Cell Culture Media
Dry powdered media for supporting cell culture growth and biomanufacturing processes.
  • Raw material components screened through strict quality control testing for lot-to-lot consistency.
  • Products are hydrated using purified process water and undergo sterile filtration.
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.
Caution: Storage 2° to 8°C, away from light