KIMAX® Specific Gravity Bottle, Hubbard, Kimble Chase
Supplier: Kimble Chase
KIMAX®
15110-24
16620-004CS 3056.85 CAD
16620-004
KIMAX® Specific Gravity Bottle, Hubbard, Kimble Chase
Specific Gravity Bottles
For specific gravity of road oils, road tars, soft tar pitches, and asphalt cements according to ASTM D70, AASHTO T43; drying oils, varnishes, resins, and related materials (ASTM D1963); water and brine (ASTM D1429); and insulating varnishes (ASTM D115)
Stopper is concave on underside and has hole, approx. 1.6 mm in diameter, to allow air to escape.