Rapid Flash Testers, Koehler
Supplier: Koehler
K16500 K16502 K16503 K16591 K16592 K16593
37000-100EA 13491.62 CAD
37000-100 37000-102 37000-104 37000-108 37000-110 37000-112
Rapid Flash Testers, Koehler
Flashpoint Testers
Units verify flash point or sustained burning qualities of small samples
Open cup models are used for determining sustained burning characteristics of mixtures of flammable and nonflammable liquids or liquids with widely different flash points when assessing flammability characteristics; temperature range is ambient to 100°C (212°F). Closed cup models are used for routine flash point testing in the range of –30 to 300°C (–22 to 572°F). Closed cup testers with an aluminum test cup have a brass lid and shutter; stainless steel test cup testers are furnished with a stainless steel lid andMore Product Information