HiVeg™ Microbiological Culture Media, HiCrome™ MRSA Agar Base to Sabouraud Maltose Broth, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
MV1151-500G MV1245-500G MV575-500G M1582-500G MV064-500G MV098-500G MV1003-500G MV1021-500G MV1023-500G MV1024-500G MV1084-500G MV1086-500G MV1139-500G MV1163-500G MV1164-500G MV1166-500G MV1213-500G MV1269-500G MV1274-500G MV133-100G MV1453-500G MV1469-500G MV1552-500G MV284-500G MV539-500G MV661-500G MV669-100G MV872-100G MV946-500G MV976-500G MV995-500G MV575-5KG MV1067-500G MV575-10KG MV1552-2.5KG MV1552-5KG
CA61000-490EA 786 CAD
CA61000-490 CA61000-592 CA61001-140 CA95021-294 CA95039-402 CA95039-404 CA95039-406 CA95039-410 CA95039-412 CA95039-414 CA95039-432 CA95039-434 CA95039-436 CA95039-442 CA95039-444 CA95039-446 CA95039-450 CA95039-456 CA95039-460 CA95039-474 CA95039-484 CA95039-492 CA95039-502 CA95039-530 CA95039-536 CA95039-540 CA95039-542 CA95039-550 CA95039-558 CA95039-564 CA95039-566 CA95040-540 CA95039-426 CA95040-542 CA89228-108 CA89228-110
HiVeg™ Microbiological Culture Media, HiCrome™ MRSA Agar Base to Sabouraud Maltose Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
The HiVeg™ line of products consists of uniquely formulated, 100% vegetable-based media designed to work in conjunction with or replace standard animal-based protein sources
The products effectively eliminate BSE/TSE contamination concerns while providing results on par with animal-based products. The wide selection of media can meet the requirements of nearly any microbiology application.