HiVeg™ Microbiological Culture Media, Selenite Cystine Fluid Medium to XLT4 Agar Base, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
MV011-500G MV093-25KG MV025-500G MV1060-500G MV107-500G MV1147-500G MV1263-500G MV1272-500G MV302-500G MV529-500G MV852-500G MV996-500G MV1255-500G
CA61000-240EA 425.3 CAD
CA61000-240 CA95024-206 CA95039-400 CA95039-424 CA95039-428 CA95039-440 CA95039-454 CA95039-458 CA95039-532 CA95039-534 CA95039-544 CA95039-568 CA95039-452
HiVeg™ Microbiological Culture Media, Selenite Cystine Fluid Medium to XLT4 Agar Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
The HiVeg™ line of products consists of uniquely formulated, 100% vegetable-based media designed to work in conjunction with or replace standard animal-based protein sources
The products effectively eliminate BSE/TSE contamination concerns while providing results on par with animal-based products. The wide selection of media can meet the requirements of nearly any microbiology application.