HiVeg™ Microbiological Culture Media, Antibiotic Assay Medium No. 36 to HiCrome™ Klebsiella Selective Agar Base, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
MV210-500G MV210-25KG MV211-25KG MV1004-500G MV1035-100G MV1035-500G MV1036-500G MV1053-500G MV1077-500G MV1146-500G MV1212-500G MV1548-500G MV1578-500G MV164-500G MV1666-500G MV1667-500G MV862-500G MV862A-500G MV893-100G MV894-100G MV971-500G
CA61000-752EA 945.31 CAD
CA61000-752 CA95024-208 CA95024-210 CA95039-408 CA95039-416 CA95039-418 CA95039-420 CA95039-422 CA95039-430 CA95039-438 CA95039-448 CA95039-500 CA95039-512 CA95039-518 CA95039-520 CA95039-522 CA95039-546 CA95039-548 CA95039-552 CA95039-554 CA95039-560
HiVeg™ Microbiological Culture Media, Antibiotic Assay Medium No. 36 to HiCrome™ Klebsiella Selective Agar Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
The HiVeg™ line of products consists of uniquely formulated, 100% vegetable-based media designed to work in conjunction with or replace standard animal-based protein sources.
  • Uniquely formulated media
  • Eliminate BSE/TSE contamination concerns
  • Meet the requirements of nearly any microbiology application
The products effectively eliminate BSE/TSE contamination concerns while providing results on par with animal-based products. The wide selection of media can meet the requirements of nearly any microbiology application.