CellPro™ Terrific Broth, Molecular Biology Grade, Sterile Filtered
Supplier: Alkali Scientific
DC0024 DC0039 DC0041
104027-830EA 72.31 CAD
104027-830 104027-100 104027-472
CellPro™ Terrific Broth, Molecular Biology Grade, Sterile Filtered
Cell Culture Media
Promote high-density bacterial growth with Alkali Scientific CellPro™ Terrific Broth, provided sterile filtered and meeting molecular biology grade standards.
  • For molecular biology
  • Liquid medium
  • 0.2 μm filtered
CellPro™ Terrific Broth is a specialized growth medium optimized for the high-density culture of Escherichia coli strains. With its enriched formulation containing tryptone, yeast extract, and glycerol, this broth supports enhanced bacterial growth and recombinant protein expression. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, providing researchers with a reliable tool for molecular biology and biotechnology applications.