CellPro™ Super Broth + Phosphates, Molecular Biology Grade, Sterile Filtered
Supplier: Alkali Scientific
DC0014 DC0023
104027-078EA 53.85 CAD
104027-078 104028-322
CellPro™ Super Broth + Phosphates, Molecular Biology Grade, Sterile Filtered
Cell Culture Media
Cultivate high-density bacterial cultures with Alkali Scientific CellPro™ Super Broth + Phosphates, provided sterile filtered and meeting molecular biology grade standards.
  • 220% more peptone and 300% more yeast extract than LB
  • Autoclave sterilized
CellPro™ Super Broth + Phosphates is a specialized growth medium optimized for the high-yield production of proteins and plasmids in Escherichia coli cultures. With its enriched formulation containing tryptone, yeast extract, and potassium phosphate, this broth supports enhanced bacterial growth and recombinant protein expression. Through meticulous sterile filtration, CellPro™ Super Broth + Phosphates maintains purity and consistency, providing researchers with a reliable tool for molecular biology and biotechnology applications.