CellPro™ Tryptone Broth, Molecular Biology Grade, Sterile Filtered
Supplier: Alkali Scientific
Prepare nutrient-rich broths for bacterial culture with Alkali Scientific CellPro™ Tryptone Broth, ensured sterile filtered and meeting molecular biology grade standards.
- For the detection of indole production by coliforms from clinical and non-clinical samples
- Sterile
- Filtered
CellPro™ Tryptone Broth is a nutrient-rich growth medium formulated for the cultivation of fastidious microorganisms. With its precise combination of tryptone, yeast extract, and sodium chloride, this broth provides essential nutrients to support microbial growth and metabolism. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, ensuring optimal performance in microbiological studies, antimicrobial susceptibility testing, and bacterial fermentation processes.