CellPro™ NZY Media, Molecular Biology Grade, Sterile Filtered
Supplier: Alkali Scientific
Ensure optimal bacterial growth with Alkali Scientific CellPro™ NZY Media, provided sterile filtered and meeting molecular biology grade standards.
- For the cultivation of recombinant E. coli and propagation of lambda bacteriophage
CellPro™ NZY Media is a nutrient-rich growth medium optimized for the cultivation of Escherichia coli strains. With its formulation of casein hydrolysate, yeast extract, and sodium chloride, this media supports rapid bacterial growth and high plasmid yield, making it ideal for molecular cloning and protein expression experiments. Through meticulous sterile filtration, CellPro™ NZY Media ensures purity and consistency, empowering researchers with reliable results in molecular biology workflows.