Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents
Supplier: Thermo Scientific
TS-63163 TS-65191 TS-65193 TS-67363
10066-392EA 534.82 CAD
10066-392 10066-396 10066-400 10066-404
Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents
Chromatography Sorbents GC Sorbents
Prepare fluoracyl derivatives for GC/MS and for flame ionization and electron-capture detection techniques with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA). These reagents react readily with alcohols, phenols, and amines.
  • Used to prepare fluoracyl derivatives for GC/MS
  • Produce stable volatile derivatives for FID and ECD techniques
  • For use withGC/MS
  • Hazards: corrosive, flammable
  • Storage: Room temperature
  • 98 to 100% standard concentration
Caution: For research use only. Not for use in diagnostic procedures.