PureClean Tyvek® 1421B Autoclave Wrappers
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
70TK01.5X01.5 70TK0202 70TK0303 70TK0404 70TK0505 70TK0606 70TK075075 70TK0808 70TK0909 70TK0911 70TK1010 70TK1212 70TK1515 70TK1530 70TK1818 70TK1822 70TK2020 70TK2424 70TK3030 70TK3636 70TK4040 70TK4080 70TK4545 70TK4848 70TK5454 70TK5472 70TK6060 70TK7272
77530-486CS 521.4 CAD
77530-486 77530-492 77530-488 77530-526 77530-522 77530-484 77530-480 77530-490 77530-496 77530-520 77530-494 77530-478 77530-504 77530-524 77530-502 77530-508 77530-472 77530-506 77530-476 77530-514 77530-498 77530-510 77530-516 77530-518 77530-512 77530-482 77530-474 77530-500
PureClean Tyvek® 1421B Autoclave Wrappers
Sterilization Wraps
PureClean™ Autoclave wrappers are made from 1421B DuPont™ Tyvek®, which is a grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
  • Uncoated Tyvek® on both sides for maximum breathability during sterilization without the chance of residue deposits on equipment post sterilization
  • Manufactured and packaged in a controlled cleanroom environment
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable