BD Vacutainer® Needle Holders, BD Biosciences
Supplier: BD Biosciences
BD Vacutainer®
364815 364815
CA-BD364815CS 140.5 CAD
VT364815 CA-BD364815
BD Vacutainer® Needle Holders, BD Biosciences
Blood Collection Tube Holders
Standard plastic holders accept disposable and reusable double-ended needles with threaded hubs
Holders convert BD Vacutainer® tube and needle into a syringe-like assembly that facilitates use. Not autoclavable.
Ordering information: To use needle holders for pediatric work, order adapter VT4597.