Utility Cart, Labconco®
Supplier: Labconco
8007000
82006-848EA 2425.3 CAD
82006-848
Utility Cart, Labconco®
Trolleys
Features two open shelves for the transport and support of small electrical equipment
Carts have a 2.5cm (1") tubular steel frame with a corrosion-resistant dry powder glacier white epoxy coating. Carts feature four 10.2cm (4") diameter casters (two with toe locks) and two handles with ergonomic extruded rubber grips. Carts include two corrosion-resistant, one-piece, vacuum-formed ABS plastic drawers to hold small labware and supplies, and a six-outlet 115V, 60Hz, 12A power strip with a cord bracket and a three-wire cord with plug. Supports up to 181kg (400lbs.).
May also be used as a compact workstation for laboratory equipment.
Ordering information: Manufacturer's lifetime warranty. Shipped fully assembled.