Accessories for 4.5 L FreeZone® Freeze Dry Systems, Labconco
Supplier: Labconco
7756200 7762800 7537800 7537801 7353403
26671-694EA 687.3 CAD
26671-694 26671-774 10148-050 10148-052 10148-064
Accessories for 4.5 L FreeZone® Freeze Dry Systems, Labconco
Freeze Dryers
Attachment port lid has a 7.62 cm (3") diameter opening to permit the use of heated drying chamber or manifolds 26617-017, -021 or -023 atop the FreeZone® 4.5 L freeze dry systems (26671-511, -513).
Wire shelf racks attach easily without tools to the sides of the cabinet to provide additional support for glassware attached to the drying chamber. Charcoal epoxy-coated steel. Each shelf measures 17.8D x 49.2W cm (7 x 193/8").