2X YT Broth, Teknova
Supplier: TEKNOVA CA
Y0220
Y0215
Y0205
Y0210
CA100218-396EA
8953.84
CAD
CA100218-396
CA100218-868
CA100219-840
CA100220-034
2X YT Broth, Teknova
Media for Microbiology
Dehydrated Media for Microbiology
2X YT is a nutritionally rich medium for cultivating recombinant strains of Escherichia coli.
- Use 31 grams per Liter to reconstitute media
- pH 7.0
Ingredients (per Liter): 1.6% Tryptone (16.0g/L), 1.0% Yeast Extract (10.0g/L), and 0.5% Sodium Chloride (NaCl) (5.0g/L). Ambient shipping conditions. Store at room temperature.