Water, CellPro™, Molecular Biology Grade, deionized (DI), autoclaved
Supplier: Alkali Scientific
DWNF1
DWNF1-20L
104028-472EA
56.92
CAD
104028-472
104028-300
Water, CellPro™, Molecular Biology Grade, deionized (DI), autoclaved
Water
CellPro™ Deionized (DI) Water represents a high-quality solution for molecular biology research. Processed through both deionization and autoclaving, this water ensures exceptional purity and sterility. With its sterile filtered formulation, CellPro™ Deionized Water provides researchers with confidence in the integrity of their experimental procedures, facilitating successful outcomes in molecular biology studies.
- For molecular biology
- Liquid Medium
- 0.2 μm filtered
Formula:
H₂O |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 UN: 3248 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...