Magnesium chloride solution 1M, CellPro™
Supplier: Alkali Scientific
DS0213
DS0189
104027-244EA
152.31
CAD
104027-244
104028-356
Magnesium chloride solution 1M, CellPro™
Magnesium chloride
CellPro™ Magnesium Chloride 1M Solution provides researchers with a high-quality source of magnesium ions for molecular biology applications. Crafted to stringent standards of purity and sterility, this solution ensures reliable performance in experimental procedures. With its sterile filtered formulation, CellPro™ Magnesium Chloride Solution offers researchers confidence and peace of mind in their work, facilitating successful outcomes in molecular biology studies.
- For molecular biology
- Liquid Medium
- 0.2 μm filtered
Formula:
MgCl₂ |
MDL Number:
MFCD00011106 CAS Number: 7786-30-3 UN: 3248 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...