D-(+)-Maltose solution 20%, CellPro™

Supplier: Alkali Scientific

DS0202 DS0185
104027-156EA 156.92 CAD
104027-156 104027-112
D-(+)-Maltose solution 20%, CellPro™
D(+)-Maltose

CellPro™ Maltose Solution stands as a trusted reagent in molecular biology research. Crafted to meticulous standards of purity and sterility, this solution offers researchers a reliable source of maltose for experimental procedures. With its sterile filtered formulation, CellPro™ Maltose Solution ensures consistent and reproducible results, empowering researchers in their molecular biology experiments.


  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered

Formula: C₁₂H₂₂O₁₁
MDL Number: MFCD00135877
CAS Number: 69-79-4
UN: 3248

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR