DrainRack™ Microscope Slide Trays, Simport Scientific
Supplier: Simport Scientific
M710-100B M710-100G M710-100P M710-100W M710-100Y
CA48648-574CS 69.87 CAD
CA48648-574 CA48648-576 CA48648-578 CA48648-580 CA48648-582
DrainRack™ Microscope Slide Trays, Simport Scientific
Slide Holders
These rugged trays/drain racks can hold up to 200 microscope slides in 100 individual numbered slots
These microscope slide trays are made of plastic.
All slides are stored upright for easier insertion and removal. Simply tilt them forward or backward with one finger to easily pick up the desired slide. Bar codes can be read without having to remove the slides from the tray. Not autoclavable.