Culture Media Bases, MilliporeSigma
Supplier: MilliporeSigma
1.01614.1000 1.02239.0500 1.02245.0500 1.03753.0500 1.03979.0500 1.04070.0500 1.05391.0500 1.07213.1000 1.07213.2500 1.07214.1000 1.07224.2500 1.07229.1000 1.08342.1000 1.08342.2500 1.10213.1000 1.11931.1000 1.11931.9025 1.15363.0500 1.01614.5000 1.07213.9025 1.03753.9025
CA1.01614.1000EA 2653.2 CAD
CA1.01614.1000 CA1.02239.0500 CA1.02245.0500 CA1.03753.0500 CA1.03979.0500 CA1.04070.0500 CA1.05391.0500 CA1.07213.1000 CA1.07213.2500 CA1.07214.1000 CA1.07224.2500 CA1.07229.1000 CA1.08342.1000 CA1.08342.2500 CA1.10213.1000 CA1.11931.1000 CA1.11931.9025 CA1.15363.0500 CA1.01614.5000 CA1.07213.9025 CA1.03753.9025
Culture Media Bases, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
This line of culture media base products is produced in powdered and granular form.
Product # 1.01614.5000 replaces and is identical to Product # 1.01614.5007.
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media. Media is granulated, except where otherwise noted.